This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07298366_g1
See other NDUFB3 GE Assays ›
Gene Symbol:
Ndufb3
Gene Name:
NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3
Gene Aliases:
2700033I16Rik, AI415450
Chromosome Location:
Chr.1: 58586599 - 58595948 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07298366_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025597.2 NP_079873.1 1 - 2 69 102
GenBank mRNA AK007930.1 - 1 - 2 68 102
AK012320.1 - 1 - 2 70 102
AK013608.1 - 1 - 2 55 102
AK138123.1 - 1 - 2 67 102
BC028669.1 - 1 - 2 225 102
BC036989.1 - 1 - 2 66 4989233 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK007930.1, AK012320.1, AK013608.1, AK138123.1, BC036989.1, NM_025597.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufb3

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
electron transport chain
oxidation-reduction process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0