This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07299618_g1
See other TNK1 GE Assays ›
Gene Symbol:
Tnk1
Gene Name:
tyrosine kinase, non-receptor, 1
Gene Aliases:
Kos1, Tnk1a, Tnk1b
Chromosome Location:
Chr.11: 69850563 - 69858730 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07299618_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnk1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031880.3 NP_114086.3 10 - 11 1580 71
GenBank mRNA AF332512.1 - 8 - 9 1400 71
AK033440.1 - 5 - 6 951 71
AK076220.1 - 10 - 11 1587 71
BC029623.1 - 9 - 10 1567 4039780 71
BC055303.1 - 9 - 10 1484 6439288 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnk1

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
phosphorylation
cell migration
peptidyl-tyrosine phosphorylation
cell differentiation
negative regulation of cell growth
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
negative regulation of Ras protein signal transduction
protein autophosphorylation
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
hormone receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0