This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07299931_m1
See other SDSL GE Assays ›
Gene Symbol:
Sdsl
Gene Name:
serine dehydratase-like
Gene Aliases:
4432411H13Rik, AI504310, SDH1, SDS-RS1, TDH
Chromosome Location:
Chr.5: 120458193 - 120472891 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07299931_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdsl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133902.2 NP_598663.2 7 - 8 859 108
GenBank mRNA AF328927.1 - 8 - 9 1126 108
AK014487.1 - 4 - 5 475 108
BC022601.1 - 7 - 8 859 4160199 108

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sdsl

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

L-serine catabolic process
threonine catabolic process
biological_process
molecular_function
L-serine ammonia-lyase activity
L-threonine ammonia-lyase activity
lyase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0