Assay Details
| Gene Symbol: Cyp4v3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133969.2 | NP_598730.1 | 8 - 9 | 1311 | 88 | |
| GenBank mRNA | AB056457.1 | - | 8 - 9 | 1277 | 88 | |
| AK004724.1 | - | 8 - 9 | 1311 | 88 | ||
| AK150653.1 | - | 8 - 9 | 1311 | 88 | ||
| AK165754.1 | - | 1278 | 88 | |||
| BC138967.1 | - | 8 - 9 | 1177 | 8861989 | 88 | |
| BC138968.1 | - | 8 - 9 | 1177 | 8861990 | 88 | |
More Information
Set Membership: |
|

Cyp4v3
Genomic Map