This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07300391_m1
See other EFEMP1 GE Assays ›
Gene Symbol:
Efemp1
Gene Name:
epidermal growth factor-containing fibulin-like extracellular matrix protein 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 28853205 - 28926743 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07300391_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Efemp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146015.2 NP_666127.2 9 - 10 1258 90
GenBank mRNA AK077302.1 - 9 - 10 1260 90
AY185605.1 - 8 - 9 1083 90
AY251056.1 - 9 - 10 1253 90
BC023060.1 - 2 - 3 214 90
BC031184.1 - 6 - 7 879 4989603 90

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Efemp1

Panther Classification:

Molecular Function -

extracellular matrix structural protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
epidermal growth factor receptor signaling pathway
peptidyl-tyrosine phosphorylation
negative regulation of chondrocyte differentiation
epidermal growth factor-activated receptor activity
epidermal growth factor receptor binding
calcium ion binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0