This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07301568_m1
See other POLR3G GE Assays ›
Gene Symbol:
Polr3g
Gene Name:
polymerase (RNA) III (DNA directed) polypeptide G
Gene Aliases:
2310047G20Rik, AV275904, RPC32, RPC7
Chromosome Location:
Chr.13: 81673837 - 81711013 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07301568_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr3g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081176.1 NP_001074645.1 5 - 6 544 63
GenBank mRNA AK009045.1 - 5 - 6 496 63
AK009876.1 - 5 - 6 498 63
AK049789.1 - 1 - 2 158 63
BC066818.1 - 5 - 6 474 30464021 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

immune system process
transcription, DNA-templated
regulation of transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
cell proliferation
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-96csp:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0