This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07301637_m1
See other RBM24 GE Assays ›
Gene Symbol:
Rbm24
Gene Name:
RNA binding motif protein 24
Gene Aliases:
6330546B05Rik, AI606861
Chromosome Location:
Chr.13: 46418180 - 46431099 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07301637_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbm24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081425.1 NP_001074894.1 1 - 2 904 59
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CJ159111.1 - 72 Chr. - - [-] on Build 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of myotube differentiation
cell differentiation
regulation of mRNA stability
nucleotide binding
nucleic acid binding
RNA binding
mRNA 3'-UTR binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0