This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07301661_g1
See other NTMT1 GE Assays ›
Gene Symbol:
Ntmt1
Gene Name:
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases:
2610205E22Rik, AL033331, AL033332, Mettl11a, NTM1A
Chromosome Location:
Chr.2: 30807780 - 30823025 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07301661_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ntmt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170592.2 NP_733480.1 2 - 3 312 77
GenBank mRNA AK011888.1 - 2 - 3 322 77
AK011960.1 - 2 - 3 314 77
AK154783.1 - 2 - 3 322 77
BC027220.1 - 2 - 3 312 3991341 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ntmt1

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid methylation
spindle organization
chromosome segregation
N-terminal peptidyl-alanine trimethylation
N-terminal peptidyl-proline dimethylation
methylation
N-terminal peptidyl-serine methylation
N-terminal peptidyl-serine dimethylation
N-terminal peptidyl-serine trimethylation
methyltransferase activity
protein methyltransferase activity
transferase activity
N-terminal protein N-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline