Assay Details
| Gene Symbol: Ceacam19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177036.5 | NP_796010.1 | 4 - 5 | 774 | 88 | |
| GenBank mRNA | AK047942.1 | - | 4 - 5 | 761 | 88 | |
| AK079290.1 | - | 4 - 5 | 775 | 88 | ||
| AK163794.1 | - | 4 - 5 | 762 | 88 | ||
| BC107227.1 | - | 4 - 5 | 702 | 40051960 | 88 | |
| BC107228.2 | - | 4 - 5 | 702 | 40051962 | 88 | |
| BC109333.3 | - | 4 - 5 | 702 | 40051723 | 88 | |
More Information
Set Membership: |
|

Ceacam19
Genomic Map