This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07302894_g1
See other TMA7 GE Assays ›
Gene Symbol:
Tma7
Gene Name:
translational machinery associated 7 homolog (S. cerevisiae)
Gene Aliases:
1110017O22Rik, Ccdc72
Chromosome Location:
Chr.9: 109077988 - 109082381 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07302894_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tma7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183250.2 NP_899073.1 3 - 4 204 77
GenBank mRNA AK002863.1 - 3 - 4 205 77
AK003762.1 - 3 - 4 205 77
AK160705.1 - 3 - 4 202 77
AK169860.1 - 3 - 4 198 77
BC034602.1 - 3 - 4 186 77
BC054363.1 - 3 - 4 177 77
BC090400.1 - 3 - 4 177 77
BC106104.1 - 3 - 4 176 5029265 77
BC116987.1 - 3 - 4 200 40126306 77
BC116989.1 - 3 - 4 200 40126308 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tma7

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0