This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07303601_g1
See other RPS24 GE Assays ›
Gene Symbol:
Rps24
Gene Name:
ribosomal protein S24
Gene Aliases:
-
Chromosome Location:
Chr.14: 24490633 - 24496146 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07303601_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207635.1 NP_997518.1 4 - 5 456 95
GenBank mRNA AK002986.1 - 819 95
BC058817.1 - 4 - 5 416 30058168 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rps24

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA processing
translation
erythrocyte homeostasis
ribosomal small subunit biogenesis
nucleotide binding
structural constituent of ribosome
translation initiation factor binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0