This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07304692_m1
See other ZBP1 GE Assays ›
Gene Symbol:
Zbp1
Gene Name:
Z-DNA binding protein 1
Gene Aliases:
2010010H03Rik, Dai, Dlm1, mZaDLM
Chromosome Location:
Chr.2: 173206612 - 173218922 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07304692_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zbp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001139519.1 NP_001132991.1 3 - 4 614 64
GenBank mRNA AK008179.1 - 3 - 4 559 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

immune system process
viral process
innate immune response
defense response to virus
positive regulation of type I interferon-mediated signaling pathway
DNA binding
left-handed Z-DNA binding
RNA binding
double-stranded RNA adenosine deaminase activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0