This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07304863_m1
See other PPM1B GE Assays ›
Gene Symbol:
Ppm1b
Gene Name:
protein phosphatase 1B, magnesium dependent, beta isoform
Gene Aliases:
PP2CB
Chromosome Location:
Chr.17: 84956741 - 85023992 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07304863_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppm1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159497.1 NP_001152969.1 1 - 2 141 89
GenBank mRNA AK076923.1 - 1 - 2 144 89
BB615769.1 - 1 - 2 142 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001159497.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
N-terminal protein myristoylation
negative regulation of interferon-beta production
peptidyl-threonine dephosphorylation
negative regulation of NF-kappaB import into nucleus
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of defense response to virus
magnesium ion binding
catalytic activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
hydrolase activity
manganese ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline