This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07304865_m1
See other H2AFY GE Assays ›
Gene Symbol:
H2afy
Gene Name:
H2A histone family, member Y
Gene Aliases:
H2AF12M, mH2a1, macroH2A1
Chromosome Location:
Chr.13: 56073622 - 56135626 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07304865_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H2afy
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159514.1 NP_001152986.1 5 - 6 821 67
NM_001159515.1 NP_001152987.1 5 - 6 818 67
GenBank mRNA AK201036.1 - 1 - 2 81 67
AK208400.1 - 1 - 2 81 67
AK209231.1 - 1 - 2 81 67
AK213934.1 - 1 - 2 81 67
AW990415.1 - 2 - 3 237 67
BC006955.1 - 5 - 6 765 2811919 67
CB574813.1 - 2 - 3 139 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
nucleosome assembly
dosage compensation
chromatin modification
regulation of lipid metabolic process
negative regulation of histone phosphorylation
regulation of gene expression, epigenetic
positive regulation of keratinocyte differentiation
negative regulation of gene expression, epigenetic
positive regulation of gene expression, epigenetic
negative regulation of histone H3-K4 methylation
negative regulation of histone H3-K27 methylation
establishment of protein localization to chromatin
negative regulation of protein serine/threonine kinase activity
negative regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
negative regulation of cell cycle G2/M phase transition
rDNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
core promoter sequence-specific DNA binding
DNA binding
chromatin binding
double-stranded methylated DNA binding
protein kinase binding
protein serine/threonine kinase inhibitor activity
chromatin DNA binding
nucleosomal DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline