This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07304887_m1
See other GJB3 GE Assays ›
Gene Symbol:
Gjb3
Gene Name:
gap junction protein, beta 3
Gene Aliases:
Cnx31, Cx31, D4Wsu144e, Gjb-3
Chromosome Location:
Chr.4: 127325235 - 127330836 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07304887_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gjb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160012.1 NP_001153484.1 1 - 2 337 114
GenBank mRNA AK083716.1 - 1 - 2 94 114
AY390400.1 - 1 - 2 345 114
CJ084102.1 - 1 - 2 342 114

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK083716.1, AY390400.1, NM_001160012.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

placenta development
cell communication
sensory perception of sound
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline