This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07304943_m1
See other TMEM9 GE Assays ›
Gene Symbol:
Tmem9
Gene Name:
transmembrane protein 9
Gene Aliases:
1500015G18Rik, AW545782
Chromosome Location:
Chr.1: 136008219 - 136035030 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07304943_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160145.1 NP_001153617.1 2 - 3 299 97
NM_001160146.1 NP_001153618.1 2 - 3 195 97
NM_025439.3 NP_079715.1 2 - 3 164 97
GenBank mRNA AK005248.1 - 2 - 3 151 97
AK021225.1 - 2 - 3 151 97
AK134909.1 - 2 - 3 129 97
AK154423.1 - 2 - 3 151 97
BC016524.1 - 2 - 3 139 3594475 97
BC020291.1 - 1 - 2 116 97
BC048017.1 - 2 - 3 118 97
BQ918002.1 - 2 - 3 386 97
CA460035.1 - 2 - 3 201 97
CX208138.1 - 2 - 3 195 97

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK005248.1, AK021225.1, AK134909.1, AK154423.1, BC016524.1, NM_001160145.1, NM_001160146.1, NM_025439.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem9

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline