This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07304944_m1
See other TMEM9 GE Assays ›
Gene Symbol:
Tmem9
Gene Name:
transmembrane protein 9
Gene Aliases:
1500015G18Rik, AW545782
Chromosome Location:
Chr.1: 136008219 - 136035030 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07304944_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160145.1 NP_001153617.1 5 - 6 637 117
NM_001160146.1 NP_001153618.1 5 - 6 533 117
NM_025439.3 NP_079715.1 5 - 6 502 117
GenBank mRNA AK005248.1 - 5 - 6 489 117
AK021225.1 - 5 - 6 489 117
AK134909.1 - 5 - 6 467 117
AK154423.1 - 5 - 6 489 117
BC016524.1 - 5 - 6 477 3594475 117
BC020291.1 - 4 - 5 454 117
BC048017.1 - 5 - 6 456 117
CA460035.1 - 5 - 6 539 117

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem9

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline