This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07305610_g1
See other FOXRED2 GE Assays ›
Gene Symbol:
Foxred2
Gene Name:
FAD-dependent oxidoreductase domain containing 2
Gene Aliases:
A430097D04Rik, D15Bwg0759e, Gm304
Chromosome Location:
Chr.15: 77940522 - 77956771 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07305610_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Foxred2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017983.2 NP_001017983.2 1 - 2 157 85
GenBank mRNA AK140027.1 - 1 - 2 117 85
BC086662.1 - 1 - 2 87 30549783 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK140027.1, BC086662.1, NM_001017983.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Foxred2

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

ER-associated ubiquitin-dependent protein catabolic process
oxidation-reduction process
glycoprotein binding
monooxygenase activity
oxidoreductase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-q595s:80/100.66.78.76:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0