This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07305616_g1
See other FOXRED2 GE Assays ›
Gene Symbol:
Foxred2
Gene Name:
FAD-dependent oxidoreductase domain containing 2
Gene Aliases:
A430097D04Rik, D15Bwg0759e, Gm304
Chromosome Location:
Chr.15: 77940522 - 77956771 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07305616_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Foxred2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017983.2 NP_001017983.2 8 - 9 1917 84
NM_001168260.1 NP_001161732.1 8 - 9 2015 84
GenBank mRNA AK089177.1 - 1 - 2 306 84
AK140027.1 - 8 - 9 1877 84
AK170795.1 - 8 - 9 1968 84
AK171308.1 - 1 - 2 306 84
BC086662.1 - 8 - 9 1847 30549783 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Foxred2

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

ER-associated ubiquitin-dependent protein catabolic process
oxidation-reduction process
glycoprotein binding
monooxygenase activity
oxidoreductase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0