This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07306539_g1
See other UQCRH GE Assays ›
Gene Symbol:
Uqcrh
Gene Name:
ubiquinol-cytochrome c reductase hinge protein
Gene Aliases:
2210416J04Rik, 2310021J10Rik, 2610041P16Rik
Chromosome Location:
Chr.4: 116066965 - 116075070 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07306539_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uqcrh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025641.3 NP_079917.1 1 - 2 156 129
GenBank mRNA AK003266.1 - 1 - 2 121 129
AK003539.1 - 1 - 2 99 129
AK011734.1 - 518 129
AK012552.1 - 1 - 2 134 129
AK018712.1 - 1 - 2 134 129
AK054005.1 - 1 - 2 122 129
AK088185.1 - 1 - 2 121 129
BC011388.1 - 1 - 2 100 3597862 129
BY138998.1 - 1 - 2 156 129
BY702808.1 - 1 - 2 129 129

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uqcrh

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, ubiquinol to cytochrome c
transport
protein heterooligomerization
oxidation-reduction process
ubiquinol-cytochrome-c reductase activity
protein complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline