This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07306625_gH
See other RPL32 GE Assays ›
Gene Symbol:
Rpl32
Gene Name:
ribosomal protein L32
Gene Aliases:
AU020185, rpL32-3A
Chromosome Location:
Chr.6: 115805514 - 115808743 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07306625_gH
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rpl32
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172086.2 NP_742083.1 2 - 3 150 99
GenBank mRNA AK002353.1 - 2 - 3 151 99
AK011017.1 - 2 - 3 151 99
AK012525.1 - 2 - 3 151 99
AK028204.1 - 2 - 3 151 99
AK146768.1 - 2 - 3 151 99
AK148453.1 - 2 - 3 151 99
AK150705.1 - 2 - 3 151 99
AK168282.1 - 2 - 3 154 99
BC030176.1 - 150 99
BC046339.1 - 2 - 3 119 6478080 99
BC086909.1 - 2 - 3 154 30122780 99

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rpl32

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline