This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07307411_s1
See other H3F3B GE Assays ›
Gene Symbol:
H3f3b
Gene Name:
H3 histone, family 3B
Gene Aliases:
9430068D06Rik, H3.3B
Chromosome Location:
Chr.11: 116021961 - 116025450 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07307411_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H3f3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008211.3 NP_032237.1 4 1514 85
GenBank mRNA AK011431.2 - 4 1512 85
AK044933.1 - 2059 85
AK146070.1 - 4 1513 85
AK151336.1 - 4 1510 85
AK151658.1 - 4 1512 85
AK151661.1 - 4 1512 85
AK151869.1 - 4 1512 85
AK152026.1 - 4 1510 85
AK152453.1 - 4 1512 85
AK152455.1 - 4 1512 85
AK152911.1 - 4 1512 85
AK153294.1 - 4 1510 85
AK153371.1 - 4 1510 85
AK159338.1 - 4 1511 85
AK159807.1 - 5 1553 85
AK162426.1 - 4 1514 85
AK163244.1 - 4 1512 85
AK166605.1 - 4 1511 85
AK167828.1 - 4 1510 85
AK168698.1 - 4 1512 85
AK168931.1 - 4 1509 85
AK169042.1 - 4 1509 85
AK169226.1 - 4 1509 85
BC002052.1 - 4 1494 85
BC021768.1 - 4 1493 3157399 85
BC037730.1 - 4 1513 4500770 85
X13605.1 - 4 1450 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene H3f3b

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
nucleus organization
muscle cell differentiation
regulation of centromere complex assembly
negative regulation of chromosome condensation
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
nucleosomal DNA binding
histone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline