This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa03225946_g1
See other NQO2 GE Assays ›
Gene Symbol:
NQO2
Gene Name:
N-ribosyldihydronicotinamide:quinone reductase 2
Gene Aliases:
-
Chromosome Location:
Chr.20: 54122293 - 54143023 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa03225946_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NQO2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027958689.1 XP_027814490.1 4 - 5 1119 101
XM_027958690.1 XP_027814491.1 7 - 8 1284 101
XM_027958691.1 XP_027814492.1 6 - 7 831 101
XM_027958692.1 XP_027814493.1 8 - 9 1148 101
XM_027958694.1 XP_027814495.1 7 - 8 1342 101
XM_027958695.1 XP_027814496.1 8 - 9 1034 101
XM_027958696.1 XP_027814497.1 8 - 9 904 101
XM_027958697.1 XP_027814498.1 7 - 8 839 101
XM_027958698.1 XP_027814499.1 7 - 8 842 101
XM_027958699.1 XP_027814500.1 6 - 7 837 101
XM_027958700.1 XP_027814501.1 8 - 9 1359 101
XM_027958701.1 XP_027814502.1 8 - 9 1362 101
XM_027958702.1 XP_027814503.1 9 - 10 1521 101
XM_027958703.1 XP_027814504.1 7 - 8 891 101
XM_027958704.1 XP_027814505.1 9 - 10 1308 101
XM_027958705.1 XP_027814506.1 9 - 10 983 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

oxidoreductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0