Assay Details
Gene Symbol: UAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977559.1 | XP_027833360.1 | 5 - 6 | 1031 | 86 | |
XM_027977565.1 | XP_027833366.1 | 6 - 7 | 1236 | 86 | ||
XM_027977570.1 | XP_027833371.1 | 5 - 6 | 1031 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
