This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa03237310_g1
See other UBE2E3 GE Assays ›
Gene Symbol:
UBE2E3
Gene Name:
ubiquitin conjugating enzyme E2 E3
Gene Aliases:
-
Chromosome Location:
Chr.2: 139068007 - 139162191 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa03237310_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2E3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_012132197.3 XP_011987587.1 3 - 4 366 99
XM_012132205.3 XP_011987595.1 3 - 4 445 99
XM_027964786.1 XP_027820587.1 3 - 4 628 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0