This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa03244542_m1
See other PEMT GE Assays ›
Gene Symbol:
PEMT
Gene Name:
phosphatidylethanolamine N-methyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.11: 28495782 - 28532141 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa03244542_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PEMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015098682.2 XP_014954168.2 4 - 5 579 59
XM_027974768.1 XP_027830569.1 4 - 5 1346 59
XM_027974769.1 XP_027830570.1 4 - 5 541 59
XM_027974770.1 XP_027830571.1 5 - 6 631 59
XM_027974771.1 XP_027830572.1 4 - 5 595 59
XM_027974773.1 XP_027830574.1 4 - 5 515 59
XM_027974774.1 XP_027830575.1 4 - 5 568 59
XM_027974775.1 XP_027830576.1 4 - 5 532 59
XM_027974776.1 XP_027830577.1 4 - 5 457 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0