Assay Details
Gene Symbol: TP53I11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016427.3 | XP_004016476.1 | 4 - 5 | 419 | 60 | |
XM_012096300.2 | XP_011951690.1 | 3 - 4 | 987 | 60 | ||
XM_027979649.1 | XP_027835450.1 | 4 - 5 | 432 | 60 | ||
XM_027979650.1 | XP_027835451.1 | 1 - 2 | 37 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
