This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa03256964_m1
See other TMEM25 GE Assays ›
Gene Symbol:
TMEM25
Gene Name:
transmembrane protein 25
Gene Aliases:
-
Chromosome Location:
Chr.15: 32015974 - 32020468 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa03256964_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_012095533.3 XP_011950923.2 3 - 4 488 57
XM_027979252.1 XP_027835053.1 2 - 3 948 57
XM_027979253.1 XP_027835054.1 2 - 3 948 57
XM_027979254.1 XP_027835055.1 3 - 4 528 57
XM_027979255.1 XP_027835056.1 3 - 4 488 57
XM_027979256.1 XP_027835057.1 3 - 4 511 57
XM_027979257.1 XP_027835058.1 3 - 4 517 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline