This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04655585_m1
See other HSD11B1 GE Assays ›
Gene Symbol:
HSD11B1
Gene Name:
hydroxysteroid 11-beta dehydrogenase 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 77036851 - 77105871 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04655585_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD11B1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009395.2 NP_001009395.1 5 - 6 723 78
XM_012186882.2 XP_012042272.1 6 - 7 946 78
XM_027975568.1 XP_027831369.1 8 - 9 1226 78
XM_027975569.1 XP_027831370.1 7 - 8 1586 78
XM_027975570.1 XP_027831371.1 7 - 8 1055 78
GenBank mRNA X69561.1 - 726 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0