This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04657452_m1
See other YAP1 GE Assays ›
Gene Symbol:
YAP1
Gene Name:
Yes1 associated transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.15: 6863369 - 7003880 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04657452_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YAP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267881.3 NP_001254810.1 7 - 8 997 61
XM_015100720.2 XP_014956206.2 8 - 9 1379 61
XM_015100721.2 XP_014956207.2 8 - 9 1374 61
XM_015100722.2 XP_014956208.2 8 - 9 1368 61
XM_015100723.2 XP_014956209.2 8 - 9 1362 61
XM_015100724.2 XP_014956210.2 7 - 8 1332 61
XM_015100725.2 XP_014956211.2 7 - 8 1326 61
XM_015100726.2 XP_014956212.2 7 - 8 1320 61
GenBank mRNA JQ714252.3 - 7 - 8 1001 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

transcription cofactor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0