This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04658581_m1
See other UCP1 GE Assays ›
Gene Symbol:
UCP1
Gene Name:
uncoupling protein 1
Gene Aliases:
UCP 1, UCP-1
Chromosome Location:
Chr.17: 18691987 - 18698653 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04658581_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001280694.1 NP_001267623.1 4 - 5 847 98
GenBank mRNA AY689125.1 - 3 - 4 369 98
JN604984.1 - 4 - 5 847 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Ovis aries

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0