This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04661191_m1
See other IL15RA GE Assays ›
Gene Symbol:
IL15RA
Gene Name:
interleukin 15 receptor subunit alpha
Gene Aliases:
-
Chromosome Location:
Chr.13: 11328954 - 11365730 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04661191_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL15RA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015099438.2 XP_014954924.2 2 - 3 472 61
XM_027976497.1 XP_027832298.1 2 - 3 472 61
XM_027976500.1 XP_027832301.1 2 - 3 471 61
XM_027976501.1 XP_027832302.1 2 - 3 471 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2tqsm:80/100.66.78.74:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0