Assay Details
| Gene Symbol: C12H1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976333.1 | XP_027832134.1 | 2 - 3 | 295 | 99 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12H1orf100| Gene Symbol: C12H1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976333.1 | XP_027832134.1 | 2 - 3 | 295 | 99 | |