Assay Details
| Gene Symbol: LOC101120058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005253.4 | XP_004005302.3 | 17 - 18 | 2051 | 75 | |
| XM_027964322.1 | XP_027820123.1 | 16 - 17 | 1979 | 75 | ||
| Gene Symbol: LOC114112956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964321.1 | XP_027820122.1 | 6 - 7 | 686 | 75 | |
More Information
Set Membership: |
|

LOC101120058
Genomic Map