Assay Details
| Gene Symbol: METTL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016361.3 | XP_004016410.1 | 4 - 5 | 670 | 56 | |
| XM_027979578.1 | XP_027835379.1 | 4 - 5 | 2777 | 56 | ||
| XM_027979579.1 | XP_027835380.1 | 4 - 5 | 2777 | 56 | ||
More Information
Set Membership: |
|

METTL15
Genomic Map