This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04766053_g1
See other MSS51 GE Assays ›
Gene Symbol:
MSS51 PPP3CB
Gene Name:
MSS51 mitochondrial translational activator
protein phosphatase 3 catalytic subunit beta
Gene Aliases:
-
-
Chromosome Location:
Chr.25: 30471467 - 30479443 on Build Oar_rambouillet_v1.0
Chromosome Location:
Chr.25: 30471467 - 30529336 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
-
Assay ID Oa04766053_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSS51
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_012105311.3 XP_011960701.1 2 - 3 332 93
Gene Symbol: PPP3CB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015104429.2 XP_014959915.1 16 - 17 5862 93
XM_015104431.2 XP_014959917.1 15 - 16 5834 93
XM_015104436.2 XP_014959922.1 14 - 15 5806 93
XM_027962274.1 XP_027818075.1 16 - 17 5865 93
XM_027962275.1 XP_027818076.1 15 - 16 5838 93
XM_027962276.1 XP_027818077.1 15 - 16 5836 93
XM_027962277.1 XP_027818078.1 14 - 15 5809 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Ovis aries

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0