This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04766055_g1
See other MSS51 GE Assays ›
Gene Symbol:
MSS51 PPP3CB
Gene Name:
MSS51 mitochondrial translational activator
protein phosphatase 3 catalytic subunit beta
Gene Aliases:
-
-
Chromosome Location:
Chr.25: 30471467 - 30479443 on Build Oar_rambouillet_v1.0
Chromosome Location:
Chr.25: 30471467 - 30529336 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
-
Assay ID Oa04766055_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSS51
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_012105311.3 XP_011960701.1 6 - 7 1277 92
Gene Symbol: PPP3CB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015104429.2 XP_014959915.1 18 - 19 7724 92
XM_015104431.2 XP_014959917.1 17 - 18 7696 92
XM_015104436.2 XP_014959922.1 16 - 17 7668 92
XM_027962274.1 XP_027818075.1 18 - 19 7727 92
XM_027962275.1 XP_027818076.1 17 - 18 7700 92
XM_027962276.1 XP_027818077.1 17 - 18 7698 92
XM_027962277.1 XP_027818078.1 16 - 17 7671 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Ovis aries

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline