This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04786372_g1
See other CERCAM GE Assays ›
Gene Symbol:
CERCAM
Gene Name:
cerebral endothelial cell adhesion molecule
Gene Aliases:
-
Chromosome Location:
Chr.3: 8149445 - 8161296 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04786372_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CERCAM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027966368.1 XP_027822169.1 12 - 13 2069 73
XM_027966369.1 XP_027822170.1 12 - 13 2027 73
XM_027966370.1 XP_027822171.1 12 - 13 3014 73
XM_027966371.1 XP_027822172.1 12 - 13 1883 73
XM_027966372.1 XP_027822173.1 12 - 13 1977 73
XM_027966373.1 XP_027822174.1 12 - 13 1841 73
XM_027966374.1 XP_027822175.1 12 - 13 1907 73
XM_027966375.1 XP_027822176.1 12 - 13 2872 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0