This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04797148_m1
See other NME6 GE Assays ›
Gene Symbol:
NME6
Gene Name:
NME/NM23 nucleoside diphosphate kinase 6
Gene Aliases:
-
Chromosome Location:
Chr.19: 53519185 - 53525809 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04797148_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NME6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004018492.4 XP_004018541.1 4 - 5 347 101
XM_012100103.3 XP_011955493.1 4 - 5 761 101
XM_012100104.3 XP_011955494.1 4 - 5 357 101
XM_012100105.3 XP_011955495.1 4 - 5 761 101
XM_012100106.3 XP_011955496.1 4 - 5 702 101
XM_012100107.3 XP_011955497.1 4 - 5 291 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0