This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04800143_g1
See other HSD11B1L GE Assays ›
Gene Symbol:
HSD11B1L
Gene Name:
hydroxysteroid 11-beta dehydrogenase 1 like
Gene Aliases:
-
Chromosome Location:
Chr.5: 17119820 - 17124825 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04800143_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD11B1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004008587.4 XP_004008636.2 7 - 8 972 81
XM_012178044.3 XP_012033434.2 7 - 8 944 81
XM_012178049.3 XP_012033439.2 6 - 7 826 81
XM_012178051.3 XP_012033441.1 6 - 7 844 81
XM_015095877.2 XP_014951363.2 6 - 7 2622 81
XM_027969818.1 XP_027825619.1 5 - 6 2504 81
XM_027969820.1 XP_027825621.1 6 - 7 903 81
XM_027969821.1 XP_027825622.1 6 - 7 813 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0