This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04805051_g1
See other TADA3 GE Assays ›
Gene Symbol:
TADA3
Gene Name:
transcriptional adaptor 3
Gene Aliases:
-
Chromosome Location:
Chr.19: 17922990 - 17933421 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04805051_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TADA3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015102307.2 XP_014957793.1 6 - 7 887 62
XM_027958096.1 XP_027813897.1 5 - 6 1270 62
XM_027958097.1 XP_027813898.1 6 - 7 887 62
XM_027958098.1 XP_027813899.1 6 - 7 884 62
XM_027958099.1 XP_027813900.1 6 - 7 884 62
XM_027958100.1 XP_027813901.1 6 - 7 1090 62
XM_027958101.1 XP_027813902.1 6 - 7 884 62
XM_027958102.1 XP_027813903.1 6 - 7 890 62
XM_027958103.1 XP_027813904.1 6 - 7 888 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0