Assay Details
| Gene Symbol: GRHPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012120962.3 | XP_011976352.1 | 7 - 8 | 851 | 66 | |
| XM_027964412.1 | XP_027820213.1 | 7 - 8 | 778 | 66 | ||
| XM_027964413.1 | XP_027820214.1 | 7 - 8 | 1057 | 66 | ||
More Information
Set Membership: |
|

GRHPR
Genomic Map