This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04818926_g1
See other RPE GE Assays ›
Gene Symbol:
RPE
Gene Name:
ribulose-5-phosphate-3-epimerase
Gene Aliases:
-
Chromosome Location:
Chr.2: 225949480 - 225977746 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04818926_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004004888.4 XP_004004937.1 4 - 5 564 82
XM_012144959.3 XP_012000349.1 5 - 6 575 82
XM_012144972.3 XP_012000362.1 4 - 5 506 82
XM_012144983.3 XP_012000373.1 4 - 5 429 82
XM_012144993.3 XP_012000383.1 5 - 6 465 82
XM_012145002.3 XP_012000392.1 5 - 6 439 82
XM_012145021.3 XP_012000411.1 4 - 5 569 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0