Assay Details
| Gene Symbol: AMDHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961436.1 | XP_027817237.1 | 9 - 10 | 1099 | 51 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AMDHD2| Gene Symbol: AMDHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961436.1 | XP_027817237.1 | 9 - 10 | 1099 | 51 | |
Set Membership: |
|