This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04823242_m1
See other GET4 GE Assays ›
Gene Symbol:
GET4
Gene Name:
guided entry of tail-anchored proteins factor 4
Gene Aliases:
-
Chromosome Location:
Chr.24: 42611853 - 42625983 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04823242_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GET4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027961264.1 XP_027817065.1 6 - 7 799 94
XM_027961265.1 XP_027817066.1 6 - 7 806 94
XM_027961266.1 XP_027817067.1 6 - 7 785 94
XM_027961267.1 XP_027817068.1 6 - 7 2573 94
XM_027961268.1 XP_027817069.1 6 - 7 2510 94
XM_027961269.1 XP_027817070.1 6 - 7 799 94
XM_027961270.1 XP_027817071.1 6 - 7 885 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0