This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04831354_g1
See other NTHL1 GE Assays ›
Gene Symbol:
NTHL1
Gene Name:
nth like DNA glycosylase 1
Gene Aliases:
-
Chromosome Location:
Chr.24: 1545632 - 1551691 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04831354_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NTHL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027961488.1 XP_027817289.1 3 - 4 933 101
XM_027961489.1 XP_027817290.1 3 - 4 531 101
XM_027961490.1 XP_027817291.1 3 - 4 540 101
XM_027961491.1 XP_027817292.1 3 - 4 1116 101
XM_027961492.1 XP_027817293.1 3 - 4 579 101
XM_027961493.1 XP_027817294.1 3 - 4 708 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline