This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04870790_m1
See other NUDT5 GE Assays ›
Gene Symbol:
NUDT5
Gene Name:
nudix hydrolase 5
Gene Aliases:
-
Chromosome Location:
Chr.13: 16913292 - 16934396 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04870790_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004014188.4 XP_004014237.1 6 - 7 590 56
XM_004014190.3 XP_004014239.1 6 - 7 615 56
XM_012188191.3 XP_012043581.1 6 - 7 594 56
XM_027976534.1 XP_027832335.1 6 - 7 668 56
XM_027976535.1 XP_027832336.1 6 - 7 671 56
XM_027976537.1 XP_027832338.1 6 - 7 670 56
XM_027976538.1 XP_027832339.1 6 - 7 671 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

pyrophosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0