Assay Details
| Gene Symbol: CRACR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967993.1 | XP_027823794.1 | 11 - 12 | 1476 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRACR2A| Gene Symbol: CRACR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967993.1 | XP_027823794.1 | 11 - 12 | 1476 | 62 | |
Set Membership: |
|