This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04894122_m1
See other PPP1R14C GE Assays ›
Gene Symbol:
PPP1R14C
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 14C
Gene Aliases:
-
Chromosome Location:
Chr.8: 81474129 - 81561303 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04894122_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R14C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027972555.1 XP_027828356.1 2 - 3 877 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t8jrk:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0