This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04897016_g1
See other CREB3L4 GE Assays ›
Gene Symbol:
CREB3L4
Gene Name:
cAMP responsive element binding protein 3 like 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 110505599 - 110511766 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04897016_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CREB3L4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_012182085.3 XP_012037475.1 7 - 8 1337 79
XM_012182126.3 XP_012037516.1 8 - 9 1188 79
XM_027975519.1 XP_027831320.1 6 - 7 1535 79
XM_027975530.1 XP_027831331.1 6 - 7 1532 79
XM_027975536.1 XP_027831337.1 7 - 8 2764 79
XM_027975539.1 XP_027831340.1 7 - 8 2761 79
XM_027975541.1 XP_027831342.1 6 - 7 1469 79
XM_027975543.1 XP_027831344.1 7 - 8 1391 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline